Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation by a broad spectrum of serine and threonine phosphatases. 1 * 20 Tablet
Fournisseur: Thermo Fisher Scientific
Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation by a broad spectrum of serine and threonine phosphatases. 1 * 20 Tablet
COMMANDE
Développer tout / Réduire tout
Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation by a broad spectrum of serine and threonine phosphatases. 1 * 20 Tablet PIERA32957 1 * 20 Tablet
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou