Protease and Phosphatase Inhibitor Mini Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation. 1 * 20 Tablet
Fournisseur: Thermo Fisher Scientific
Protease and Phosphatase Inhibitor Mini Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation. 1 * 20 Tablet
COMMANDE
Développer tout / Réduire tout
Protease and Phosphatase Inhibitor Mini Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation. 1 * 20 Tablet PIERA32959 1 * 20 Tablet
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou