Protease inhibitor tablets, EDTA-free, Pierce™
Fournisseur: Thermo Fisher Scientific
A32965
PIERA32965EA 378 EUR InStock
PIERA32965
Protease inhibitor tablets, EDTA-free, Pierce™
Suppléments et additifs pour culture cellulaire
These inhibitor tablets contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments.
  • Formulated without EDTA, a metalloproteinase inhibitor
  • Ready-to-use and fully disclosed
  • Can be used directly with Pierce BCA assays
  • Improved formulation helps the tablets dissolve quickly to form a clear solution
  • Do not contain phosphatase inhibitors
Protease inhibitor tablets protect proteins from proteolytic degradation through inhibition of the serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin, and pepstatin A.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité