Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate), No-Weigh™ Format, Pierce™

Fournisseur: Thermo Fisher Scientific
Attention

A35394
PIERA35394EA 157 EUR InStock
PIERA35394
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate), No-Weigh™ Format, Pierce™
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Thermo Scientific Pierce SMCC is a hetero-bifunctional crosslinker that contain N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules. NHS esters react with primary amines at pH 7–9 to form amide bonds, while maleimides react with sulfhydryl groups at pH 6.5–7.5 to form stable thioether bonds. In aqueous solutions, NHS ester hydrolytic degradation is a competing reaction whose rate increases with pH. The maleimide group is more stable than the NHS-ester group, but will slowly hydrolyze and lose its reaction specificity for sulfhydryls at pH values > 7.5. For these reasons, conjugations with these crosslinkers are usually performed at pH 7.2–7.5, with the NHS ester (amine-targeted) reacted before or simultaneous with the maleimide (sulfhydryl-targeted) reaction.

  • Reactive groups: NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-SMCC
  • High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
  • Cyclohexane bridge confers added stability to the maleimide group making SMCC an ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate (SMCC) is a non-cleavable and membrane permeable crosslinker. It contains an amine-reactive N -hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of SMCC and Sulfo-SMCC and are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm.

Attention: For Research Use Only. Not for use in diagnostic procedures.


Formule: C₁₆H₁₈N₂O₆
Point de fusion: 179 °C
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00009634
Numéro CAS: 64987-85-5
Commander maintenant
Résultats des tests de spécification
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO

 

COMMANDE

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité