Slide-A-Lyzer™ G3 Dialysis Cassettes
Fournisseur: Thermo Fisher Scientific
Slide-A-Lyzer™
514-1389EA 169 EUR InStock
514-1389 514-1400 514-1401 514-1402 514-1385 514-1386 514-1387 514-1388 514-1381 514-1382 514-1383 514-1384 514-1380 514-1378 514-1379 514-1396 514-1397 514-1398 514-1399 514-1392 514-1393 514-1394 514-1395 514-1390 514-1391
Slide-A-Lyzer™ G3 Dialysis Cassettes
Tuyaux Membranes de dialyse
Thermo Scientific Slide-A-Lyzer™ G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 0,5 to 125 ml. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.
  • Efficient dialysis - flat cassette chamber, housing two membranes, provides high surface area to volume ratio maximising diffusion rate
  • Ease-of-use - sample loading and removal with a pipette and syringe
  • High sample recovery - low binding plastic and membrane help minimise sample loss compared to filtration and resin systems
  • Maximum sample security - sealed membranes prevent leakage that can occur with dialysis tubing and homemade devices, caps have visual and haptic feedback assuring secure closure of the sample chamber
  • Stream-lined design - less plastic, lighter weight devices
  • Self-floating format - designed to float independently without the use of external buoys
  • Irradiated option - E-beam irradiated cassettes are available for applications requiring conditions that minimise risk of sample contamination
The Slide-A-Lyzer G3 Cassettes are single-use dialysis devices designed to maximise performance and practicality. They provide an easy-to-use and secure alternative to traditional dialysis tubing and are used in a wide range of applications including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.
The G3 cassettes are ergonomically designed to be significantly lighter and more compact than the previous generations, making them easier to handle and more practical to use. This design offers a new cap allowing effortless sample loading and retrieval providing visual and haptic feedback upon opening and closure to assure sample transfer and security. Sample loading and removal is easily accomplished using a serological pipette or hypodermic needle (optional) attached to a syringe. All G3 cassettes offer a built-in air chamber that provides buoyancy and vertical orientation during the dialysis process.
Slide-A-Lyzer G3 Dialysis Cassettes are available in five different sizes, accommodating sample volumes ranging from 0,5 to 125 ml, and are provided with 2K, 3,5K, 10K, and 20K molecular weight cut offs (MWCOs). The cassettes membranes are composed of low-binding regenerated cellulose that provides maximum sample purity and recovery. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.
The Slide-A-Lyzer G3 Cassettes with the 10K MWCO are also offered in the Electron Beam treated format for applications such as culturing cells, purifying viruses, or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. A dosage that has been determined to sterilize units was used based on bioburden testing for identifying anaerobic growth. Sterility testing is not performed on individual lots.
Membrane: Cellulose membrane
Purification target: Buffer exchange, protein
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité