Wound and eyewash sprays
Fournisseur: Plum Safety
4554
PLUM4554EA 28 EUR InStock
PLUM4554
Wound and eyewash sprays
Matériel de premiers secours Produits de premiers secours
This spray provides immediate and effective care to minor accidents such as cuts and scrapes on the skin or dust and dirt in the eyes. It helps to minimises the risk of further infection or damage.
  • Quick and easy single hand operation
  • Safe, precise and effective rinsing
  • Dust cap to protect the spray nozzle from contamination
  • Easy to transport, classed as a non flammable aerosol
  • Environmental friendly and fully recyclable
  • Hypoallergenic
  • Sterile solution without preservatives
It is a portable, easy to use, saline-based and pain-free spray. It works at any angle, providing a balanced flow rate of sterile 0,9% saline solution, strong enough for washing away clot and foreign bodies from wounds, yet soft enough for washing the eyes.
Ideal for the industrial sector, production industry, laboratories, transport of dangerous goods, first responders, law enforcement, firefighters, military and first rescue services, such as disaster relief.
Shelf-life: 3 years. Expiry date is shown on the bottom of the can.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité