Plum Pull1Aid® Wall First Aid Kit
Fournisseur: Plum Safety
5181
PLUM5181EA 266 EUR InStock
PLUM5181
Plum Pull1Aid® Wall First Aid Kit
Matériel de premiers secours Matériel d'urgence
Plum Pull1Aid® is a wall mounted first aid station that fits most workplaces. A flexible solution that gives a quick overview of the available first aid and makes it easy to always have the right products in the right place.
  • Clear cover - hygienic and protects the products from dust
  • Practical and functional station
  • High quality products
  • Simple to use with clear instructions
  • Easy and quick to refill
  • Immediate and visible access to plasters, ready to use with a pull
  • Wound care products are quick to pull out of the stations
Pull1Aid® allows you to combine first aid and wound care products that suit the needs at your workplace. The flexible combinations provide a tailored solution and avoid excess products.
Pull1Aid® is clear, fast and simple first aid so you can act immidiately if an accident happens. Expand your Pull1Aid® station for increased safety for wounds and eyes by mounting a QuickSafe® station next to it.
Kit includes:
1 × QuickCool burn gel
1 × QuickClean wound cleanser
1 × QuickStop wound pressure dressing
2 × Blood stopper wound pressure dressing
1 × Blood stopper mini wound pressure dressing
1 × Burn gel dressing for burns
1 × Protect kit respiratory aid
1 × 200 ml wound and eyewash spray
1 × QuickFix elastic
1 × QuickRinse
QuickRinse eye ampoules for quick rinsing in the event of a foreign body getting into the eye. The solution gently rinses the eye and prevents further contamination. QuickCool burn gel with tea tree oil provides a cooling and pain-relieving effect. The QuickStop dressing set contains 3 dressings: one large one for stopping, for example, arterial hemorrhage and two smaller ones for use in minor bleeding. In addition to cleaning wounds, Wound and Eyewash Spray can also be used to cleanse the eyes of foreign bodies. Blood Stoper 4 in 1 dressings stop bleeding. The mini version is perfect for dressing the smallest wounds, example on hands and fingers. Burn Gel is a sterile gel dressing that cools and relieves pain from burns. The Protect Kit respiration kit enables safe resuscitation and effectively protects against the victim's secretions.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité