22° Bevel matched point needle sets
Fournisseur: CADENCE SCIENCE
4372
POPR4372EA 0 EUR InStock
POPR4372
22° Bevel matched point needle sets
Aiguilles pour seringues
These needle sets are manufactured to strict standards providing assurance of perfectly matched bevels. They incorporate type 304 stainless steel needles and stylets to assure needle point strength, sharpness, and integrity of point geometry. The nickel plated brass hubs and stylet caps have matching serial numbers to facilitate matching after cleaning or sterilisation. The point of the needle is aligned with the flat of the hub. The hub slot and cap guide are aligned with the point of the needle for easier orientation.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité