Mussel tissue - organics, NIST-2974A standard
Fournisseur: LGC Standards PROMOCHEM
NIST-2974A
PROONIST-2974AEA 2310 EUR InStock
PROONIST-2974A
Mussel tissue - organics, NIST-2974A standard
Étalons Étalons organiques
This standard reference material is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, polybrominated diphenyl ether (PBDE) congeners, hexabromocyclododecane (HBCD) isomers, methylmercury, inorganic mercury and total mercury in marine bivalve mollusk tissue and similar matrices.
A unit consists of one bottle containing approximately 5 g of freeze-dried mussel tissue.
Certifications: Traceable to NIST and SRM numbers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité