Standards for water pollution QC check, hexavalent chromium, VHG
Fournisseur: LGC Standards PROMOCHEM
VHG-QWPCR6-15
PROOVHG-QWPCR6-15EA 209 EUR InStock
PROOVHG-QWPCR6-15
Standards for water pollution QC check, hexavalent chromium, VHG
Étalons Étalons pour l'analyse environnementale
This solution is intended for use as a quality control check for methods related to hexavalent chromium.
This standard has been prepared as a concentrate and must be diluted prior to analysis and should be analysed as soon as possible after the concentrate is diluted. This concentrate is not preserved but can be stored at room temperature.
Certifications: Traceable to NIST SRM.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité