Standards for water pollution QC check, inorganics, VHG
Fournisseur: LGC Standards PROMOCHEM
VHG-QWSIN-500
PROOVHG-QWSIN-500EA 270 EUR InStock
PROOVHG-QWSIN-500
Standards for water pollution QC check, inorganics, VHG
Étalons Étalons pour l'analyse environnementale
This solution is intended for use as a quality control check for methods related to measurements of inorganic materials includes conductivity, alkalinity as CaCO₃, chloride, fluoride, nitrite and nitrate, sulphate, potassium, sodium, total dissolved solids and many more.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
Certifications: Traceable to NIST SRM.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité