Standards for water pollution QC check, residual chlorine, VHG
Fournisseur: LGC Standards PROMOCHEM
VHG-QWSRCL-2
PROOVHG-QWSRCL-2EA 152 EUR InStock
PROOVHG-QWSRCL-2
Standards for water pollution QC check, residual chlorine, VHG
Étalons Étalons pour l'analyse environnementale
This solution is intended for use as a quality control check for methods related to free and total residual chlorines.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is not preserved. Since the diluted standard is not stable, it must be analysed immediately after the concentrate is diluted.
Certifications: Traceable to NIST SRM.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité