Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CEACAM5 Mouse Monoclonal Antibody [clone: C66/261]

Fournisseur: ProSci Inc.
33-110
PRSI33-110EA 773 EUR InStock
PRSI33-110
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: C66/261]
Anticorps
This antibody recognises proteins of 80-200 kDa, identified as different members of the CEA (Carcinoembryonic Antigen) family. CEA is synthesised during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This antibody does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. The antibody shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+). CEA antibody positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.

ELISA: order BSA-free format for coating (2) Flow Cytometry: 0,5-1 µg/million cells IF: 1 - 2 µg/ml IHC (FFPE): 1 - 2 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (3) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate Buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. Clone C66/261 (recommended capture) will pair with clone C66/1009 (recommended detect) by ELISA. It will NOT pair with clone C66/1030.
3. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: C66/261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité