Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TG Mouse Monoclonal Antibody [clone: 2H11 or TGB04]

Fournisseur: ProSci Inc.
33-209
PRSI33-209EA 773 EUR InStock
PRSI33-209
Anti-TG Mouse Monoclonal Antibody [clone: 2H11 or TGB04]
Anticorps
Thyroglobulin is a 660 kDa dimeric pre-protein with mutiple glycosylation sites, detected at ~300 kDa in western blot. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for thyroglobulin antibody even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently thyroglobulin antibody negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Thyroglobulin antibody, combined with calcitonin antibody, can identify medullary carcinomas of the thyroid. Furthermore, thyroglobulin antibody, combined with TTF1 antibody, can be a reliable marker to differentiate between primary thyroid and lung neoplasms.

Flow Cytometry: 0,5-1 µg/million cells IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: 2H11 or TGB04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité