3913P
PRSI3913PEA
122
EUR
InStock
PRSI3913P
Avian Influenza hemagglutinin 4 Influenza hemagglutinin 4 Peptide (10 aa in the center)
Protéines et Peptides
Avian influenza hemagglutinin 2 peptide is used for blocking activity of avian influenza hemagglutinin 2 antibody. It usually blocks antibody activity completely in ELISA assays by incubating peptide with equal volume of antibody for 30 min at 37 °C antibody.