3925P
PRSI3925PEA
122
EUR
InStock
PRSI3925P
Avian Influenza hemagglutinin 1 Influenza hemagglutinin 1 Peptide (14 aa in the center)
Protéines et Peptides
Avian influenza hemagglutinin 2 peptide is used for blocking activity of avian influenza hemagglutinin 2 antibody. It usually blocks antibody activity completely in ELISA assays by incubating peptide with equal volume of antibody for 30 min at 37 °C antibody.