Human recombinant Activin A (from E. coli)
Fournisseur: ProSci Inc.
40-213
PRSI40-213EA 305 EUR InStock
PRSI40-213
Human recombinant Activin A (from E. coli)
Protéines et Peptides
Activin A is a TGF-β family member that exhibits a wide range of biological activities including regulation of cellular proliferation and differentiation, and promotion of neuronal survival. Elevated levels of Activin A in human colorectal tumors and in post-menopausal woman have been implicated in colorectal and breast cancers, respectively. The biological activities of Activin A can be neutralized by inhibins and by the diffusible TGF-β antagonist, Follistatin. Human Activin A is a 26.0 kDa disulfide-linked homodimer of two β A chains, each containing 116 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité