Human Recombinant NT4 (from E. coli)
Fournisseur: ProSci Inc.
40-566
PRSI40-566EA 461 EUR InStock
PRSI40-566
Human Recombinant NT4 (from E. coli)
Protéines et Peptides
NT-4 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-3. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-4 is expressed in the prostate, thymus, placenta and skeletal muscle. NT-4 can signal through the LNGFR and trkB receptors and promotes the survival of peripheral sensory sympathetic neurons. Recombinant human NT-4 is a noncovalently linked homodimer, of two 14.0 kDa polypeptide monomers (260 total amino acid residues). Manufactured using all non-animal reagents.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité