Human recombinant IL1B (fromE. coli)
Fournisseur: ProSci Inc.
40-598
PRSI40-598EA 461 EUR InStock
PRSI40-598
Human recombinant IL1B (fromE. coli)
Protéines et Peptides
IL-1β is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1α and IL-1β binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1β is predominantly a cell-associated cytokine. Recombinant human IL-1β is a 17.3 kDa protein containing 153 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité