7343P
PRSI7343PEA
122
EUR
InStock
PRSI7343P
Human DIS3 peptide
Protéines et Peptides
18 amino acids peptide near the amino terminus of human DIS3.
- Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide