90-006
PRSI90-006EA
654
EUR
InStock
PRSI90-006
Mouse recombinant Resistin (from HEK293 cells)
Protéines et Peptides
The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELM-alpha/FIZZ 1 and RELM-beta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.