90-215
PRSI90-215EA
448
EUR
InStock
PRSI90-215
Human recombinant FasL (soluble) (from HEK293 cells)
Protéines et Peptides
FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. Is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.