Mouse recombinant BTNL2 (from CHO cells)
Fournisseur: ProSci Inc.
90-375
PRSI90-375EA 686 EUR InStock
PRSI90-375
Mouse recombinant BTNL2 (from CHO cells)
Protéines et Peptides
Butyrophilin-like 2 (BTNL2) is a butyrophilin family member with homology to the B7 costimulatory molecules, polymorphisms of which have been recently associated through genetic analysis; to sporadic inclusion body myositis and sarcoidosis. BTNL2 mRNA is highly expressed in lymphoid tissues as well as in intestine. BTNL2-Ig fusion protein was shown to recognize a putative receptor whose expression on B and T cells was significantly enhanced after activation. It inhibited T cell proliferation and TCR activation of NFAT, NF-kappaB and AP-1 signalling pathways. BTNL2 is the first member of the butyrophilin family that regulates T cell activation, which has implications in immune diseases and immunotherapy.
  • This recombinant proteins is for research use only.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité