Mouse recombinant IL12 (from CHO cells)
Fournisseur: ProSci Inc.
90-503
PRSI90-503EA 686 EUR InStock
PRSI90-503
Mouse recombinant IL12 (from CHO cells)
Protéines et Peptides
Interleukin-12 (IL-12), also known as Natural Killer Cell Stimulatory Factor (NKSF) or Cytotoxic Lymphocyte Maturation Factor (CLMF), is a heterodimeric pleiotropic cytokine made up of a 40 kDa (p40) subunit and a 35 kDa (p35) subunit. IL-12 is produced by macrophages and B lymphocytes and has been shown to have multiple effects on T cells and Natural Killer (NK) cells. Some of these IL-12 activities include the induction of IFN-gamma and TNF in resting and activated T and NK cells; the enhancement of cytotoxic activity of resting NK and T cells, the stimulation of resting T cell proliferation in the presence of a comitogen; and the enhancement of NK cell proliferation. Current evidence indicates that IL-12 is a key mediator of cellular immunity and induces the differentiation of Th1 cells from precursor T helper cells. Based on its activities, it has been suggested that IL-12 may have therapeutic potential as a vaccine adjuvant that promotes cellular immunity and as an antitumour and anti viral agent.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité