91-374
PRSI91-374EA
884
EUR
InStock
PRSI91-374
Human recombinant Kallikrein 5 (from cells)
Protéines et Peptides
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many Kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen Kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag