91-453
PRSI91-453EA
654
EUR
InStock
PRSI91-453
Human recombinant SUMF1 (from cells)
Protéines et Peptides
Human Sulfatase Modifying Factor 1 (SUMF1) is a 42kDa protein. SUMF1 is a Ca2+-binging member of the sulfatase-modifying factor family. SUMF1 is a soluble ER lumenal glycoprotein, it converts inactive sulfatases into an active form by transforming a catalytic site cysteine into a formylglycine residue. In the ER, SUMF1 can exist as either a monomer, or a disulfide-linked homodimer or a heterodimer with SUMF2. Three splice isoforms are known.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag