91-640
PRSI91-640EA
654
EUR
InStock
PRSI91-640
Human recombinant FGF8B (from E. coli)
Protéines et Peptides
Fibroblast growth factor 8 (FGF8) is a member of the fibroblast growth factor family. It is discovered as a growth factor essential for the androgen-dependent growth of mouse mammary carcinoma cells. Mouse FGF8b shares 100% aa identity with human FGF8b. FGF8 is widely expressed during embryogenesis, and mediates epithelial-mesenchymal transitions. It plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. It is required for normal brain, eye, ear, limb development during embryogenesis and normal development of the gonadotropin-releasing hormone (GnRH) neuronal system.
- This recombinant protein can be used for biological assays. For research use only.