Human Recombinant Galectin 3 (from Cells)
Fournisseur: ProSci Inc.
91-660
PRSI91-660EA 588 EUR InStock
PRSI91-660
Human Recombinant Galectin 3 (from Cells)
Protéines et Peptides
Galectin-3(LGALS3) is also known as Galactose-specific lectin 3, Mac-2 antigen, Carbohydrate-binding protein 35, Laminin-binding protein and Galactoside-binding protein. LGALS3 is highly expressed in early stages of papillary carcinoma, and lowly during tumor progression. LGALS3 is probably forms homo- or heterodimers and secreted by a non-classical secretory pathway and associates with the cell surface. LGALS3 plays an important role during the acquisition of vasculogenic mimicry and angiogenic properties. LGLAS3 takes part in an immune regulator to inhibit T-cell immune responses and promote tumor growth, as a result providing a new mechanism for tumor immune tolerance.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité