Human Recombinant High Mobility Group B3 (from Cells)
Fournisseur: ProSci Inc.
91-678
PRSI91-678EA 654 EUR InStock
PRSI91-678
Human Recombinant High Mobility Group B3 (from Cells)
Protéines et Peptides
High Mobility Group Protein B3 (HMGB3) belongs to the HMGB family. Members of the HMG box subfamily are thought to be have an important role in DNA replication, nucleosome assembly and transcription. HMGB3 binds preferentiallly single-stranded DNA and unwinds double stranded DNA. HMGB3 consists of 200 amino acids and is localized to the cell nucleus. It contains two HMG box DNA-binding domain. HMGB3 binds preferentially single-stranded DNA and unwinds double stranded DNA.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité