Human recombinant INSL4 (from cells)
Fournisseur: ProSci Inc.
91-712
PRSI91-712EA 852 EUR InStock
PRSI91-712
Human recombinant INSL4 (from cells)
Protéines et Peptides
Early Placenta Insulin-Like Peptide (INSL4) belongs to the insulin family. INSL4 is expressed in the early placental cytotrophoblast and syncytiotrophoblast INSL4 is a secreted protein and a precursor that undergoes post-translational cleavage to produce 3 polypeptide chains, A-C, that form tertiary structures composed of either all three chains, or just the A and B chains. INSL4 plays an important role in the development of trophoblast and in the regulation of bone formation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité