Human recombinant Reg3 gamma (from cells)
Fournisseur: ProSci Inc.
91-855
PRSI91-855EA 852 EUR InStock
PRSI91-855
Human recombinant Reg3 gamma (from cells)
Protéines et Peptides
Pancreatitis-associated protein IB, Regenerating islet-derived protein III-gamma, is a secreted protein which contains 1 C-type lectin domain. It is expressed almost exclusively in the pancreas and also expressed in testis, but not found in small intestine. This protein might be a stress protein involved in the control of bacterial proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité