91-855
PRSI91-855EA
852
EUR
InStock
PRSI91-855
Human recombinant Reg3 gamma (from cells)
Protéines et Peptides
Pancreatitis-associated protein IB, Regenerating islet-derived protein III-gamma, is a secreted protein which contains 1 C-type lectin domain. It is expressed almost exclusively in the pancreas and also expressed in testis, but not found in small intestine. This protein might be a stress protein involved in the control of bacterial proliferation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag