Human recombinant CEACAM21 (from cells)
Fournisseur: ProSci Inc.
91-865
PRSI91-865EA 852 EUR InStock
PRSI91-865
Human recombinant CEACAM21 (from cells)
Protéines et Peptides
Carcinoembryonic antigen-related cell adhesion molecule 21 is a protein that in humans is encoded by the CEACAM21 gene. It belongs to the immunoglobulin superfamily. CEA family.containing 1 Ig-like C2-type (immunoglobulin-like) domain.It was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité