91-874
PRSI91-874EA
620
EUR
InStock
PRSI91-874
Human recombinant CXCL8 (from cells)
Protéines et Peptides
Interleukin-8 (IL-8) belongs to the neutrophil-specific CXC family of chemokines. It is one of the initial cytokines released from a variety of cell types, including T cells, endothelial cells and fibroblasts, in response to an inflammatory stimulus and acts by recruiting neutrophils, T-cells and basophils to the site of inflammation. Elevated Interleukin-8 levels are associated with the onset of a variety of disease states.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag