Human recombinant IL2 (from cells)
Fournisseur: ProSci Inc.
91-937
PRSI91-937EA 409 EUR InStock
PRSI91-937
Human recombinant IL2 (from cells)
Protéines et Peptides
Interleukin-2(IL-2) is an interleukin, a type of cytokine signaling molecule in the immune system,belongs to the IL-2 family. It is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité