91-971
PRSI91-971EA
884
EUR
InStock
PRSI91-971
Human recombinant PBLD (from E. coli)
Protéines et Peptides
Phenazine Biosynthesis-Like Domain-Containing protein (PBLD) belongs to the phenazine biosynthesis-like protein (PhzF) family, which is expressed in most tissues. PBLD takes part in the MAPK signaling pathway, and is involved in multiple basic cellular functions. The expression of PBLD can be increased in several disease processes, including insulin resistance, folate deficiency and hypotension.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag