Human recombinant PBLD (from E. coli)
Fournisseur: ProSci Inc.
91-971
PRSI91-971EA 884 EUR InStock
PRSI91-971
Human recombinant PBLD (from E. coli)
Protéines et Peptides
Phenazine Biosynthesis-Like Domain-Containing protein (PBLD) belongs to the phenazine biosynthesis-like protein (PhzF) family, which is expressed in most tissues. PBLD takes part in the MAPK signaling pathway, and is involved in multiple basic cellular functions. The expression of PBLD can be increased in several disease processes, including insulin resistance, folate deficiency and hypotension.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité