Human recombinant UBE2J2 (from E. coli)
Fournisseur: ProSci Inc.
91-996
PRSI91-996EA 884 EUR InStock
PRSI91-996
Human recombinant UBE2J2 (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 J2 (UBE2J2) belongs to the ubiquitin-conjugating enzyme family. UBE2J2 is involved in the ubiquitiantion. UBE2J2 located in the membrane of the endoplasmic reticulum, catalyzes the covalent attachment of ubiquitin to other proteins. UBE2J2 may play a important role in the selective degradation of misfolded membrane protein from the endoplasmic reticulum.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité