92-049
PRSI92-049EA
654
EUR
InStock
PRSI92-049
Human Recombinant Galectin-Related (from E. coli)
Protéines et Peptides
Galectin-Related Protein (LGALSL) is a 172 amino acid protein that contains one Galectin domain. LGALSL does not appear to bind carbohydrates or lactose as the critical residues required for binding are not conserved. LGALSL may play a significant role in stimulating smooth muscle growth in developing alveolar wall vessels and the development of pulmonary capillaries.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag