92-466
PRSI92-466EA
852
EUR
InStock
PRSI92-466
Human recombinant KLRB1 (from cells)
Protéines et Peptides
Killer cell lectin-like receptor subfamily B, member 1(KLRB1) is a single-pass type II membrane protein which contains 1 C-type lectin domain. KLRB1 plays an inhibitory role on natural killer (NK) cells cytotoxicity. Activation results in specific acid sphingomyelinase/SMPD1 stimulation with subsequent marked elevation of intracellular ceramide. Activation also leads to AKT1/PKB and RPS6KA1/RSK1 kinases stimulation as well as markedly enhanced T-cell proliferation induced by anti-CD3. It acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope as well as to the N-acetyllactosamine epitope. Binds also to CLEC2D/LLT1 as a ligand and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.