Mouse recombinant GMCSF (from E. coli)
Fournisseur: ProSci Inc.
92-499
PRSI92-499EA 852 EUR InStock
PRSI92-499
Mouse recombinant GMCSF (from E. coli)
Protéines et Peptides
Granulocyte-macrophage colony-stimulating factor is an enzyme that in mouse is encoded by the Csf2 gene, belongs to the GM-CSF family.CSF2 is a Cytokine that stimulates the growth and differentiation of hematopoietic precursor cells from various lineages, including granulocytes, macrophages, eosinophils and erythrocytes
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité