Human Recombinant HOXB4 (from E. coli)
Fournisseur: ProSci Inc.
92-525
PRSI92-525EA 760 EUR InStock
PRSI92-525
Human Recombinant HOXB4 (from E. coli)
Protéines et Peptides
Homeobox B4 (HOXB4) is encoded by the HOXB4 gene which is a member of the the class I homeobox (HOX) gene family and encodes a nuclear protein with a homeobox DNA-binding domain. These genes are master control regulators of developmental programs including embryonic and adult hematopoiesis. Multiple HOX genes, including HOXB4, are highly expressed in the hematopoietic stem cells (HSC) compartment. HOXB4 gene can act in opposite ways when expressed by different cells, promoting the proliferation of stem cells whilst activating the apoptotic pathway in some embryonic structures. The protein HOXB4, as a homeodomain transcription factor, has been shown to be an important regulator of stem cell renewal and hematopoiesis. Incellular or ectopic expression of HOXB4 expands hematopoietic stem and progenitor cells in vivo and in vitro, making it a potential candidate for therapeutic stem cell expansion.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité