92-531
PRSI92-531EA
852
EUR
InStock
PRSI92-531
Mouse recombinant TGF beta 2
Protéines et Peptides
Transforming growth factor beta 2 (TGF- beta 2) is a member of TGF-beta superfamily that shares a characteristic cysteine knot structure. Mice with TGF- beta 2 gene deletion show defects in development of cardiac, lung, craniofacial, limb, spinal column, eye, inner ear and urogenital systems. All TGF- beta isoforms signal via the same heteromeric receptor complex, consisting of a ligand binding TGF- beta receptor type II (T beta R-II), and a TGF- beta receptor type I (T beta R-I). Signal transduction from the receptor to the nucleus is mediated via SMADs. TGF- beta expression is found in cartilage, bone, teeth, muscle, heart, blood vessels, haematopoitic cells, lung, kidney, gut, liver, eye, ear, skin, and the nervous system.
- This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.