92-571
PRSI92-571EA
588
EUR
InStock
PRSI92-571
Mouse recombinant IL7RA
Protéines et Peptides
Interleukin 7 Receptor alpha (IL-7R alpha), also known as CD127, is a 75 kDa hematopoietic receptor superfamily member that plays an important role in lymphocyte differentiation, proliferation, and survival. IL-7R alpha is majorly expressed on T cells and their precursors, and early in B cell development as well, prior to the appearance of surface IgM. Dynamic regulation of IL-7R alpha is important for the generation of appropriate immune responses.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.