92-614
PRSI92-614EA
654
EUR
InStock
PRSI92-614
Human recombinant CTGF (from cells)
Protéines et Peptides
CTGF belongs to the CCN (CTGF/Cyr61/Cef10/NOVH) protein family, which is comprised of six secreted proteins that reside in the extracellular matrix (ECM). CTGF causes a variety of cellular responses including reduced cell adhesion and enhanced cell migration and proliferation. CTGF has also been shown to be essential for epithelial to mesenchymal transition (EMT), a process whereby normal functioning cells morph into ones that produce mainly scar tissue (of which collagen in the major protein component).
This recombinant protein can be used for biological assays. For research use only.