Human recombinant CTGF (from cells)
Fournisseur: ProSci Inc.
92-614
PRSI92-614EA 654 EUR InStock
PRSI92-614
Human recombinant CTGF (from cells)
Protéines et Peptides
CTGF belongs to the CCN (CTGF/Cyr61/Cef10/NOVH) protein family, which is comprised of six secreted proteins that reside in the extracellular matrix (ECM). CTGF causes a variety of cellular responses including reduced cell adhesion and enhanced cell migration and proliferation. CTGF has also been shown to be essential for epithelial to mesenchymal transition (EMT), a process whereby normal functioning cells morph into ones that produce mainly scar tissue (of which collagen in the major protein component).
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité