Mouse recombinant MBL2
Fournisseur: ProSci Inc.
92-681
PRSI92-681EA 588 EUR InStock
PRSI92-681
Mouse recombinant MBL2
Protéines et Peptides
Mannose-binding Lectin (MBL) is an acute phase protein bearing to the family of collectins produced by the liver as a monomer that forms a triple helix. Once released in serum, it further polymerises forming dimers to octamers. The degree of serum polymerisation is critical for the biological activity of MBL. MBL has higher affinity to microbial polysaccharides or their glycoconjugates. MBL was shown earlier to bind cell surfaces of bacteria, fungi, protozoa and viruses and acts as an acute-phase plasma protein (APP) during infection and inflammation. MBL activates the lectin-complement pathway, promotes opsonophagocytosis and modulates inflammation.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité