96-065
PRSI96-065EA
448
EUR
InStock
PRSI96-065
Human recombinant BMF2 (from E. coli)
Protéines et Peptides
Human bone morphogenetic protein 2 (BMP2) is also known as human BMP-2, BMP-2, BMP2, BMP 2, h-BMP-2, rh-BMP-2, recombinant human BMP-2, recombinant BMP-2, BMP, is a member of the BMP subgroup belonging to the TGF-b superfamily of structurally related signaling proteins. BMP-2 is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with osteoconductive carriers such as collagen and synthetic hydroxyapatite. In addition to its osteogenic activity, BMP-2 plays an important role in cardiac morphogenesis and is expressed in a variety of tissues including lung, spleen, brain, liver, prostate ovary and small intestine. As implied by its name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. BMP2 has been demonstrated to potently induce osteoblast differentiation in a variety of cell types, and induce apoptosis in human myeloma cell lines as a novel function.