96-636
PRSI96-636EA
727
EUR
InStock
PRSI96-636
Human recombinant Epcr (from HEK293 cells)
Protéines et Peptides
Endothelial protein C receptor (PROCR) is also known as Activated protein C receptor (APC receptor), Endothelial cell protein C receptor, CD antigen CD201. PROCR / EPCR / CD201 is a N-glycosylated protein, with a soluble form exists. PROCR is probably released by a metalloprotease. PROCR seems to have the same activity as the membrane-bound form. PROCR / EPCR / CD201 is expressed strongly in the endothelial cells of arteries and veins in heart and lung. PROCR binds activated protein C and enhances protein C activation by the thrombin - thrombomodulin complex. PROCR plays a role in the protein C pathway controlling blood coagulation.