HCASMC, Cryopreserved
Fournisseur: PROVITRO
1210612
PRTV1210612EA 1191 EUR InStock
PRTV1210612
HCASMC, Cryopreserved
Cellules
HCASMC - Human coronary artery smooth muscle cells - are obtained from human tissue, in particularly from the coronary artery smooth muscle cells, initially isolated in their in vivo state. The cells are then transitioned into a primary cell culture and subsequently aliquoted and frozen for preservation.
  • It's crucial to note that HKER cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
  • Each batch of HKER undergoes rigorous testing, specifically assessing their proliferative capacity. These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HKER cultures for research and experimental applications.
It's crucial to note that HCASMC cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
Each batch of HCASMC undergoes rigorous testing, specifically assessing their proliferative capacity.
These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HCASMC cultures for research and experimental applications. This product contain 500,000 cells/cryovial.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HCASMC samples are shipped on dry ice, they must be immediately transferred to a freezer at –80 °C for storage, either for a period of up to one week or after the thawing process described below.
In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HCASMC samples for longer-term storage requirements.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HKER samples are shipped on dry ice, they must be immediately transferred to a freezer at −80 °C for storage, either for a period of up to one week or after the thawing process described below. In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HKER samples for longer-term storage requirements.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité