Fibroblast Supplement-Mix, FCS
Fournisseur: PROVITRO
2170401
PRTV2170401EA 91 EUR InStock
PRTV2170401
Fibroblast Supplement-Mix, FCS
Suppléments et additifs pour culture cellulaire
The supplement kit for defined fibroblast maintenance medium is suitable for maintenance culturing of Provitro human fibroblasts (HFIB) after adding the supplement kit components to 500 ml basal medium. The formulation is optimized for initial seeding of 6000 cells/cm². Feeder-layer, matrix substrates or other substances are not necessary. We recommend using the antibiotic supplement for freshly isolated cells only.
Storage: Samples remain stable for a duration of twelve months from the receipt date when stored within the temperature range of −20 to −80 °C. To ensure optimal storage conditions, maintain a sterile environment at these temperatures. For improved preservation, it is advisable to aliquot the protein. Avoid subjecting the samples to repeated freeze-thaw cycles to prevent degradation and maintain the integrity of the material.
Attention: Store the supplements at −20 °C. Do not use the supplements after expiry date. Avoid any thaw/freeze cycle.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité