Arctic Winter Gloves
Fournisseur: Portwest
A146YERM
PRTWA146YERMEA 7.6 EUR InStock
PRTWA146YERM
Arctic Winter Gloves
Gants
Twin liner traps in heat, and a ³/₄ nitrile sandy dipping offers greater protection to the fingers as well as improved liquid protection. Open back for breathability. A fantastic glove for a wide range of industries.
  • Specially designed for use in cold conditions
  • Twin liner traps in heat through increased insulation
  • Provides contact heat protection up to 250 °C for 15 seconds
  • Flexible sandy nitrile coating offers great grip in wet and dry conditions
  • ³/₄ dipped for increased protection
  • Superb abrasion and tear resistance
  • 15 gauge liner for extra dexterity
  • OEKO-TEX® STANDARD 100 approved
Low linting construction for minimal contamination, with a breathable seamless liner. Retail tag aids presentation for retail sales.
Colour: Yellow
Certifications: EN ISO 21420: 2020 Dexterity 3, EN 388: 2016 + A1: 2018 4242X, EN 407: 2020 X2XXXX, EN 511: 2006 X2X. ANSI/ISEA 105: 2016 CUT Level A7, ANSI/ISEA 105: 2016 ABRASION Level 4. CE certified.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité