Weatherproof Hi-Vis Gloves
Fournisseur: Portwest
A450ORRXL
PRTWA450ORRXLEA 73.5 EUR InStock
PRTWA450ORRXL
Weatherproof Hi-Vis Gloves
Gants
The A450 is designed for use in the most demanding conditions. A tough double-dipped PVC coating prevents oil, grease, and water penetration.
  • Fully coated for maximum liquid protection
  • Double-dipped coating for durability
  • Prevents grease, oil, and water penetration
  • 12 gauge liner for dexterity
  • Knitted cuff for comfort and warmth
Suitable for any work area where oil and grease resistance is a priority, these gloves feature an insulated liner and knit wrist to keep hands warm and dry.
Colour: Orange
Certifications: EN ISO 21420: 2020, EN 388: 2016 + A1: 2018 3121X, EN 511: 2006 11X.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité